WIN-55,212-2 ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:131543-23-2
:
Formula:C₂₇H₂₆N₂O₃·CH₃SO₃H
:522.6 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity:≥98% (HPLC)
Specification Test Results
Frequently Bought Together