Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
A-803467
A-803467 ≥98%
A-803467 ≥98%
Catalog # ENZOBMLNA1420050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
A-803467 ≥98%
Catalog # ENZOBMLNA1420050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
50 mg
Seq:
2370
Specifications
About this item
Product Details
:
:
357.79 g/mol
CAS Number:
944261-79-4
Formula:
C₁₉H₁₆ClNO₄
MDL Number:
MFCD10574689
Product Details