
Sodium nitroprusside dihydrate
Catalog # PROOBP453
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:200 mg
- Seq:2530
Specifications
About this item
:
Merck Index:13,08722
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1588
ADR:6.1,III
:297.95 g/mol
EINECS:238-373-9
:GHS06
CAS Number:13755-38-9
Formula:Na₂[Fe(CN)₅NO]·2H₂O
MDL Number:MFCD00149192