0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
Enzymes
Pepsin
Pepsin
MOLEM13218112
:
Molekula
:
Pepsin
MOLEM13218112
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
100 g
CAS No.:
9001-75-6
Storage conditions:
Refrigerator
Enzyme name:
Pepsin