
Sodium nitroprusside dihydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13755-38-9
:
Formula:Na₂[Fe(CN)₅NO]·2H₂O
MDL Number:MFCD00149192
Merck Index:14,8649
Density:1.72
Storage Temperature:Ambient
EINECS:238-373-9
UN:1588
ADR:6.1,III
:297.95 g/mol
:GHS06
Related Information
Frequently Bought Together