0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
TLC Development Tanks
Accessories for Twin Trough Chambers
Accessories for Twin Trough Chambers
Accessories for Twin Trough Chambers
Catalog # CAMA022.5292
Supplier:
CAMAG
Expand image
Accessories for Twin Trough Chambers
Catalog # CAMA022.5292
Supplier:
CAMAG
Specifications
Description:
Glass lid for CAMAG® Twin trough chamber made from DURAN® Borosilicate 3.3 for 20×10 cm plates
Pk:
1
Specifications
About this item
Product Details
Product Details