Order Entry
Hungary
ContactUsLinkComponent
Native Human Concanavalin A (from Human), Alexa Fluor® 594
Native Human Concanavalin A (from Human), Alexa Fluor® 594
Catalog # AATB25590
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Human Concanavalin A (from Human), Alexa Fluor® 594
Catalog # AATB25590
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Alexa Fluor® 594
  • Protein/peptide type:
    Native
  • Species:
    Human
  • Protein synonyms:
    ConA
  • Protein/peptide name:
    Concanavalin A

Specifications

About this item

Concanavalin A (ConA) is a lectin that binds specifically to certain structures found in various sugars, glycoproteins and glycolipids.

  • Well-cited conjugate
  • High affinity and selectivity
  • Multiple applications
Caution: For research use only.