Amplite® IR
Catalog # AATB11009
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:1 mg
- Description:Amplite™ IR
Specifications
About this item
Our Amplite® IR is a fluorogenic peroxidase substrate that generates near infrared fluorescence upon reaction with peroxidase and H2O2.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.