Order Entry
Hungary
ContactUsLinkComponent
Amplite® Colorimetric Glucose Oxidase Assay Kit
Amplite® Colorimetric Glucose Oxidase Assay Kit
Catalog # AATB11299
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Colorimetric Glucose Oxidase Assay Kit
Catalog # AATB11299
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    500 Tests
  • Description:
    Amplite™ colorimetric glucose oxidase assay kit
Specifications
About this item
The glucose oxidase is a dimeric protein that catalyses the oxidation of beta-D-glucose into hydrogen peroxide and D-glucono-1,5-lactone, which is hydrolysed to gluconic acid.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.