Amplite® Colorimetric Sphingomyelinase Assay Kit *Blue Color*
Catalog # AATB13620
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pk:200 Tests
- Description:Amplite™ colorimetric sphingomyelinase assay kit *blue color*
Specifications
About this item
Sphingomyelinase (SMase) is an enzyme that is responsible for cleaving sphingomyelin (SM) to phosphocholine and ceramide.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.