Rendelés felvitel
Hungary
ContactUsLinkComponent
Amplite® Fluorimetric Acetylcholine Assay Kit *Red Fluorescence*
Amplite® Fluorimetric Acetylcholine Assay Kit *Red Fluorescence*
Catalog # AATB11403
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Fluorimetric Acetylcholine Assay Kit *Red Fluorescence*
Catalog # AATB11403
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Kiszerelés:
    200 Tests
  • Leírás:
    Amplite™ fluorimetric acetylcholine assay kit *red fluorescence*
Specifications
About this item
Acetylcholine and its metabolites are needed for three main physiological purposes: structural integrity and signaling roles for cell membranes, cholinergic neurotransmission (acetylcholine synthesis), and as a major source for methyl groups via its metabolite, trimethylglycine (betaine) that participates in the S-adenosylmethionine synthesis pathways.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.