Concanavalin A, XFD594
AATB25575
:
- Pack type:Vial
- Pk:1 mg
- Conjugation:Alexa Fluor® 594
- Protein/peptide type:Native
- Species:Human
- Protein synonyms:ConA
- Protein/peptide name:Concanavalin A
XFD594 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 594 (Alexa Fluor® is the trademark of ThermoFisher).
- Well-cited conjugate
- High affinity and selectivity
- Multiple applications
: For research use only.