Native Human Wheat Germ Agglutinin (from Eukaryotic cells), Alexa Fluor® 555
AATB25539
:
- Pack type:Vial
- Pk:1 mg
- Conjugation:Alexa Fluor® 555
- Protein/peptide type:Native
- Source:Eukaryotic cells
- Species:Human
- Protein synonyms:WGA
- Protein/peptide name:Wheat Germ Agglutinin
Wheat germ agglutinin (WGA) is a lectin that binds to N-acetyl-D-glucosamine and sialic acid.
- Glycoconjugate binding
- Very bright signal
- Multiple applications
: For research use only.