Order Entry
Hungary
ContactUsLinkComponent
 
Native Streptavidin (from Streptomyces avidinii), iFluor® 488
 
undefined
Native Streptavidin (from Streptomyces avidinii), iFluor® 488

 

  • Conjugation:
    iFluor® 488
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin|Streptomyces avidinii
  • Protein/peptide name:
    Streptavidin

 

Frequently Bought Together