BSB [(trans,trans)-1-Bromo-2,5-bis-(3-hydroxycarbonyl-4-hydroxy)styrylbenzene]
Catalog # AATB23050
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:481.292 g/mol
CAS Number:56776-28-4
Formula:C₂₄H₁₇BrO₆