
EGTA-Na4 (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt) 10 mM calcium chelator
Catalog # AATB21008
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:468.28 g/mol
ADR:5,II
:GHS07
CAS Number:13368-13-3
Formula:C₁₄H₂₀N₂Na₄O₁₀
MDL Number:MFCD00274420