Order Entry
Hungary
ContactUsLinkComponent
5-Propargylamino-3'-azidomethyl-dCTP
Irritant
5-Propargylamino-3'-azidomethyl-dCTP
Catalog # AATB17091
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
5-Propargylamino-3'-azidomethyl-dCTP
Catalog # AATB17091
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 Nmo
  • Seq:
    9030

Specifications

About this item

 :
 :575.26 g/mol
ADR:5,II
 :GHS07
CAS Number: 666847-71-8
Formula:C13H20N7O13P3