Order Entry
Hungary
ContactUsLinkComponent
Cyanine 3 hydrazide ≥95% [equivalent to Cy3® hydrazide]
Cyanine 3 hydrazide ≥95% [equivalent to Cy3® hydrazide]
Catalog # AATB146
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cyanine 3 hydrazide ≥95% [equivalent to Cy3® hydrazide]
Catalog # AATB146
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :872.85 g/mol