Recombinant Human FCGRT/B2M (from HEK293 cells), His-tag
Catalog # ABNOP9771.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His-tag
- Gene ID:P55899-1 | P61769-1
- Protein/peptide name:FCGRT/B2M
- Sequence:Ala24-Ser297|Ile21-Met119