Order Entry
Hungary
ContactUsLinkComponent
 
Recombinant Human LEP (from E. coli)
Catalog # ABNOP8893.50UG
Supplier:  Abnova
CAS Number:  
Recombinant Human LEP (from E. coli)
Catalog # ABNOP8893.50UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    P41159
  • Protein/peptide name:
    LEP

Specifications

About this item