Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human TNFRSF1A (from CHO cells)
Recombinant Human TNFRSF1A (from CHO cells)
Recombinant Human TNFRSF1A (from CHO cells)
ABNOP7363.10UG
:
Abnova
:
Recombinant Human TNFRSF1A (from CHO cells)
ABNOP7363.10UG
:
Abnova
:
Pk:
10 µG
Protein/peptide type:
Recombinant
Source:
CHO cells
Species:
Human
Gene ID:
P19438
Protein/peptide name:
TNFRSF1A
Sequence:
50a.a.-211a.a