Recombinant Human CCL20 (from E. coli)
Catalog # ABNOP7475.5UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:P78556
- Protein/peptide name:CCL20
- Sequence:27a.a.-96a.a.