Recombinant Human CXCL12 (from E. coli)
Catalog # ABNOP7203.10UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:P48061
- Protein/peptide name:CXCL12
- Sequence:22a.a.-89a.a.