Recombinant Mouse CXCL5 (from E. coli)
Catalog # ABNOP7226.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Gene ID:P50228
- Protein/peptide name:CXCL5
- Sequence:41a.a.-132a.a.