Anti-FAS Rabbit Recombinant Antibody
Catalog # ABNORAB02496.100UL
Supplier: Abnova
Specifications
- Antigen symbol:FAS
- Clonality:Recombinant
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human FAS.
- Pk:100 µl