0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
Antibodies
Anti-ENO1 Rabbit Recombinant Antibody
Anti-ENO1 Rabbit Recombinant Antibody
ABNORAB02460.100UL
:
Abnova
Anti-ENO1 Rabbit Recombinant Antibody
ABNORAB02460.100UL
:
Abnova
Antigen symbol:
ENO1
Clonality:
Recombinant
Host:
Rabbit
Reactivity:
Human
Immunogen:
Recombinant rabbit monoclonal antibody raised against human ENO1.
Pk:
100 µl