Anti-Pollen allergen (BETVIA) Rabbit Recombinant Antibody [clone: BV16]
Catalog # ABNORAB03700.200UG
Supplier: Abnova
Specifications
- Antigen symbol:Pollen allergen (BETVIA)
- Clonality:Recombinant
- Clone:BV16
- Host:Rabbit
- Reactivity:Birch (Betula pendula)
- Immunogen:Recombinant rabbit monoclonal antibody raised against Birch pollen allergen (BETVIA).
- Pk:200 µG