Order Entry
Hungary
ContactUsLinkComponent
Anti-PRDM9 Rabbit Recombinant Antibody [clone: RAB-C370]
Anti-PRDM9 Rabbit Recombinant Antibody [clone: RAB-C370]
Catalog # ABNORAB03763.200UG
Supplier:  Abnova
Anti-PRDM9 Rabbit Recombinant Antibody [clone: RAB-C370]
Catalog # ABNORAB03763.200UG
Supplier:  Abnova

Specifications

  • Antigen symbol:
    PRDM9
  • Clonality:
    Recombinant
  • Clone:
    RAB-C370
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human PRDM9.
  • Pk:
    200 µG

Specifications

About this item