Anti-PRMT3 Human Recombinant Antibody [clone: RAB-C372]
ABNORAB03769.100UG
: Abnova
- Antigen symbol:PRMT3
- Clonality:Recombinant
- Clone:RAB-C372
- Host:Human
- Reactivity:Human
- Immunogen:Recombinant human monoclonal antibody raised against human PRMT3.
- Pk:100 µG