Anti-TBP Rabbit Recombinant Antibody [clone: 3C7]
ABNORAB03973.100UL
: Abnova
- Antigen symbol:TBP
- Clonality:Recombinant
- Clone:3C7
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TBP.
- Pk:100 µl