Order Entry
Hungary
ContactUsLinkComponent
Anti-FH Rabbit Recombinant Antibody [clone: R04-5B6]
Anti-FH Rabbit Recombinant Antibody [clone: R04-5B6]
Catalog # ABNORAB02280.100UL
Supplier:  Abnova
Anti-FH Rabbit Recombinant Antibody [clone: R04-5B6]
Catalog # ABNORAB02280.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    FH
  • Clonality:
    Recombinant
  • Clone:
    R04-5B6
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human FH.
  • Pk:
    100 µl

Specifications

About this item