Experimentation kits, blood typing
Catalog # EDVO140
Supplier: EDVOTEK
Specifications
- Description:Blood Typing
- Pk:1
Specifications
About this item
ABO and Rh typing of blood left at the scene of a crime can help to narrow down a list of suspects. In this experiment, students use agglutination to identify the blood group of unknown blood samples as a step to identify a criminal.
- For 10 lab groups
- Complete in 45 minutes
Also required: Microscope with 400x magnification.
Delivery: Kit includes instructions, control ABO Rh simulated blood samples, unknown simulated blood samples, transfer pipettes, and microscope slides.