Accessories for Stationary Tank System for Lubricants, MULTI-Tank
Catalog # CEMG11452
Gyártó: CEMO
Specifikációk
- Description:Electronic overfill prevention, comprises evaluation unit and probe housing protection category IP 54, fitting 25,4 mm and 19 mm, reduction 50,8 mm to 25,4 mm, probe 500 mm, 230 VAC
- Pk:1