Order Entry
Hungary
ContactUsLinkComponent
Stationary Tank System for Lubricants, MULTI-Tank
Stationary Tank System for Lubricants, MULTI-Tank
Catalog # CEMG7536
Gyártó:  CEMO
Stationary Tank System for Lubricants, MULTI-Tank
Catalog # CEMG7536
Gyártó:  CEMO
Specifikációk
  • Capacity:
    400 L
  • Description:
    MULTI-Tank stationary tank system, HDPE
  • W×D×H:
    740×730×1170 mm
  • Pk:
    1
Specifikációk
Termék információk
Tank from HDPE with general construction inspection approval with rigidly connected sheet steel pallet for handling with a forklift or pallet truck from any of the four sides.
  • Tank as described above with integral sheet steel pallet for driving under with a forklift or hand forklift from any of four sides
  • Approved for transport according to the ADR, but without a pump fitted
  • Lower centre of gravity increases stability and improves handling/ transport
  • For storage of diesel, biodiesel without a bund, even in water-protected areas
  • Certified for installation indoors
Individual tank with integral galvanised sheet steel bund, standard accessories such as filling contents gauge, optical leakage detector device, integral carrying handle, no pump.