Order Entry
Hungary
ContactUsLinkComponent
Molykote® High Vaccum Grease
Molykote® High Vaccum Grease
Catalog # DOWC23900246
Supplier:  DOW CORNING
Molykote® High Vaccum Grease
Catalog # DOWC23900246
Supplier:  DOW CORNING
Specifications
  • Description:
    High vaccum grease
  • Pack type:
    Tube
  • Pk:
    50 g
Specifications
About this item
Molykote® High-Vacuum grease features a stiff, non-melting silicone formula that helps to seal and lubricate chemical processing equipment. Suitable for use in a wide range of applications including sealing vacuum and pressure systems, it is also doubles as a lubricant for o-rings in binoculars and telescopes as it prevents fogging of delicate lenses.
  • Helps to reduce wear in plastic-to-metal and rubber-to-metal contact
  • Low volatility for applications involving chemical processing equipment
  • Excellent resistance to water, chemicals, high and low temperatures
  • Helps prevent fogging of delicate lenses
Applications: Lubrication, sealing, chemical processing equipment, O-rings, vaccum and pressure systems.
Certifications: Meets FDA 21 CFR 175.300 and NSF 61.