Order Entry
Hungary
ContactUsLinkComponent
Scotch-Weld™ DP804 EPX Super Clear Acrylic Adhesive
Scotch-Weld™ DP804 EPX Super Clear Acrylic Adhesive
Catalog # MMMADP804
Supplier:  3M
undefined
Scotch-Weld™ DP804 EPX Super Clear Acrylic Adhesive
Catalog # MMMADP804
Supplier:  3M
Specifications
  • Volume:
    48,5 ml
  • Colour:
    Transparent
  • Description:
    Scotch-Weld™ EPX Super clear acrylic adhesive DP804
  • Pk:
    12
Specifications
About this item
The Scotch-Weld™ EPX™ Bonding system is a discharging system for mixing two-component mixed adhesives. The main agent and the curing agent with the cartridge are mixed and discharged by a special hand gun and a special nozzle.
  • Easy mixing and application with the help of the EPX gun
  • 3 minutes of working time/4 minutes of handling time
  • Exposure at temperatures from 0 to 100 °C
  • Easy to apply
Stable adhesion can be obtained by eliminating the hassle of troublesome six-liquid mixing.