Order Entry
Hungary
ContactUsLinkComponent
Amersham™ ECL Gold Hybridization Buffer, Cytiva
Amersham™ ECL Gold Hybridization Buffer, Cytiva
Catalog # RPN3006
Supplier:  Cytiva
CAS Number:  
undefined
Amersham™ ECL Gold Hybridization Buffer, Cytiva
Catalog # RPN3006
Supplier:  Cytiva
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    ECL Gold Hybridization Buffer
  • Application:
    Sufficient for 4000 cm² Membrane

Specifications

About this item

ECL Direct Nucleic Acid Labeling and Detection Systems are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction. The resulting probe can be used without purification. Detection is achieved by generation of light via the HRP-catalyzed breakdown of luminol.

  • Direct probe labelling in a 10 minute reaction, 1 hour from hybridization to detection with ECL Direct and Hybond™ N+.
  • Eliminates handling, waste and regulatory issues associated with use of radioactivity.
  • No need to strip blots before reprobing.
  • Contains a novel rate enhancer, giving higher sensitivity with very low backgrounds on nylon membranes.
Delivery: ECL Direct is particularly useful for applications that require a blot to be reprobed several times and also for screening applications requiring rapid throughput and fast results. Each system includes the following reagents, sufficient for labeling 5 to 10 µg nucleic acid and detecting 2000 to 4000 cm ² of membrane (depending on product ordered): labeling reagent, crosslinker, control DNA, blocking agent, ECL Detection Reagents, and ECL Gold Hybridization Buffer.