Order Entry
Hungary
ContactUsLinkComponent
Cole-Parmer® Essentials Bottle Top Dispensers with Recirculation Valve
Cole-Parmer® Essentials Bottle Top Dispensers with Recirculation Valve
Catalog # STUA25208-31
Supplier:  Cole Parmer
Cole-Parmer® Essentials Bottle Top Dispensers with Recirculation Valve
Catalog # STUA25208-31
Supplier:  Cole Parmer

Specifications

  • Volume:
    0,5 - 5 ml
  • Description:
    Bottle-top dispenser with recirculation valve
  • Seq:
    01
  • Accuracy:
    ±0,6 %
  • Division:
    0,1 ml
  • Imprecision:
    ±0,2 %
  • Pk:
    1

Specifications

About this item

Save money by conserving reagents.

  • Recirculation valve prevents loss of reagent during priming and purging
  • Smooth plunger movement provides bubble-free dispensing
  • Adjustable nozzle allows users to change the dispensing angle
  • Wetted materials: PTFE, ETP, borosilicate glass, PP, PFA, FEP
  • Autoclavable

These bottle top dispensers conserve reagents with a built-in recirculation valve, allowing users to prime and purge without waste. The PTFE piston with ETP O-ring provide frictionless movement, reducing the occurrence of bubbles in dispenses. The springless PTFE valve manifold offers superior chemical compatibility and resists jamming.

Delivery: Includes discharge tube, telescoping filling tube, calibration tool, bottle adapter set (28, 32, 38, 40, and 45 mm) and calibration certificate.