Order Entry
Hungary
Orders LinkContactUsLinkComponent
Pimobendan, Reference Standards

Pimobendan, Reference Standards

Supplier:   USP
Pimobendan, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1569315.
  • 1569326.
  • 1569337.
  • Description:
  • Pimobendan
  • Pimobendan Related Compound A
  • Pimobendan Related Compound B
  • Synonym(s):
  • 4-[2-(4-Methoxyphenyl)-1H-benzimidazol-5-yl]-3-methyl-4-oxobutanoic acid
  • N-[2-Amino-4-(4-methyl-6-oxo-1,4,5,6-tetrahydropyridazin-3-yl)phenyl]-4-methoxybenzamide
  • CAS No.:
  • 74150-27-9
  • 74149-74-9
  • UN No.:
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1569315.
    Pimobendan
    Pimobendan
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 382 000 Ft
    382 000 Ft
    1 * 100 mg
       382 000 Ft

    Specifications:
    More
    Specifications
    Cat. No.
    1569315.
    Description
    Pimobendan
    Synonym(s)
    CAS No.
    74150-27-9
    UN No.
  • 1569326.
    Pimobendan Related Compound A
    Pimobendan Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 436 000 Ft
    436 000 Ft
    1 * 15 mg
       436 000 Ft

    Specifications:
    More
    Specifications
    Cat. No.
    1569326.
    Description
    Pimobendan Related Compound A
    Synonym(s)
    4-[2-(4-Methoxyphenyl)-1H-benzimidazol-5-yl]-3-methyl-4-oxobutanoic acid
    CAS No.
    UN No.
    2811
  • 1569337.
    Pimobendan Related Compound B
    Pimobendan Related Compound B
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 458 000 Ft
    458 000 Ft
    1 * 15 mg
       458 000 Ft

    Specifications:
    More
    Specifications
    Cat. No.
    1569337.
    Description
    Pimobendan Related Compound B
    Synonym(s)
    N-[2-Amino-4-(4-methyl-6-oxo-1,4,5,6-tetrahydropyridazin-3-yl)phenyl]-4-methoxybenzamide
    CAS No.
    74149-74-9
    UN No.