Order Entry
Hungary
ContactUsLinkComponent
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)
Irritant
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)
Catalog # BC15
Supplier:  G-Biosciences
CAS Number:  
undefined
Irritant
EMCH.HCl (E-Maleimidocaproic acid hydrazine hydrochloride)
Catalog # BC15
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Spacer Arm (Å):
    11.8
  • Reactive Toward:
    Sulfhydryl + Carbohydrate
  • Membrane Permeable:
    Not tested
  • Water Soluble:
    No
  • Cleavable/ Reversible:
    No

Specification Test Results

About this item

 :
 :261.705 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:175290-73-0
 :151038-94-7
Formula:C₁₀H₁₅N₃O₃·ClH
MDL Number:MFCD07363434