Order Entry
Hungary
ContactUsLinkComponent
Anti-RAB5A Rabbit polyclonal antibody
Anti-RAB5A Rabbit polyclonal antibody
Catalog # NOVUNB120-13253
Supplier:  Novus Biologicals
Anti-RAB5A Rabbit polyclonal antibody
Catalog # NOVUNB120-13253
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ras-related protein Rab-5
  • Antigen symbol:
    Rab5a
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    5868
  • Antigen synonyms:
    RAB5RAS-associated protein RAB5A|RAB5A|ras-related protein Rab-5A|member RAS oncogene family
  • Storage buffer:
    PBS and 50% Glycerol
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    The antibody recognizes 26kDa Rab5 of the above species origins.Human Rab5 synthetic peptide conjugated to KLH; identical to dog Rab5 sequence over the residues
  • Purification:
    Protein A purified
  • Pk:
    0,1 mg

Specifications

About this item

The Rab5a Antibody from Novus Biologicals is a rabbit polyclonal antibody to Rab5a. This antibody reacts with human, mouse. The Rab5a Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Rab5a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse