Order Entry
Hungary
ContactUsLinkComponent
Anti-TBX2 Rabbit Polyclonal Antibody
Anti-TBX2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-89459
Supplier:  Novus Biologicals
Anti-TBX2 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-89459
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TBX2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Gene ID:
    6909
  • Antigen synonyms:
    FLJ10169|T-box protein 2|T-box 2|T-box transcription factor TBX2
  • Storage buffer:
    PBS (pH 7,2) and 40% Glycerol
  • Storage temperature:
    Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    This antibody was developed against Recombinant Protein corresponding to amino acids:IPMPTFGGLFPYPYTYMAAAAAAASALPATSAAAAAAAAAGSLSRSPFLGSARPRLRFSPYQIPVTIPPSTSLLTTGLASEGSKAAGGNSREPSPLPELALRKVGAPSRGALSPSGSAKEAANELQSIQRLVSGL
  • Purification:
    Immunogen affinity purified
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

The TBX2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TBX2. This antibody reacts with human, mouse. The TBX2 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TBX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse