Rendelés felvitel
Hungary
ContactUsLinkComponent
PBA plates, Bond Elut
PBA plates, Bond Elut
Catalog # VARI7541201C
Supplier:  VARIAN
PBA plates, Bond Elut
Catalog # VARI7541201C
Supplier:  VARIAN
Specifications
  • Description:
    VersaPlate preassembled 96-well plate, PBA, 100 mg (96 wells)
  • Pk:
    1
Specifications
About this item
Unique bond Elut PBA silica sorbent uses phenylboronic acid functionality to retain analytes via a reversible covalent bond, with high specificity and cleanliness.
  • Unique phenylboronic acid sorbent
  • High specificity for cis-diol compounds
  • Amenable for a broad range of biomolecule applications
Bond Elut PBA is a unique silica SPE sorbent containing a phenylboronic acid functionality that can retain analytes through a reversible covalent bond. This very strong covalent retention mechanism enables high specificity and cleanliness.
The boronate group has a strong affinity for cis-diol containing compounds, such as catechols, nucleic acids, some proteins, carbohydrates, and PEG compounds. Amino alcohols, alpha-hydroxy amides, keto compounds, and others can also be retained.