Order Entry
Hungary
ContactUsLinkComponent
SPME fibers
SPME fibers
  VARISU57326U
 :  VARIAN
SPME fibers
  VARISU57326U
 :  VARIAN
  • Description:
    SPME fiber, DVB/PDMS, manual, 65 µm, Stableflex
  • Pk:
    1
SPME fibers are available in a range of chemistries, formats, and for use with autosamplers or manual injections.
  • SPME fibers can be used multiple times when treated with the proper care and caution
  • Each fiber has a colour-coded or notched hub, indicating the type of coating on the fiber
Kits are also available to support method development.
 : When ordering SPME fibers, note that the fiber kits contain fibers only. For a first-time order, you will also need to order the appropriate fiber holder for your needs.