Order Entry
Hungary
ContactUsLinkComponent
PPL Tubes, Bond Elut™
  VARI7531901C
 :  VARIAN
PPL Tubes, Bond Elut™
  VARI7531901C
 :  VARIAN
  • Description:
    VersaPlate well plate tubes, PPL, 100 mg
  • Pk:
    24
Bond Elut PPL (Priority PolLutant) is a modified styrene-divinylbenzene polymer that retains polar analytes, including phenols, for excellent absolute recoveries.
  • Modified styrene-divinylbenzene polymer
  • Large particle size allows fast extraction speeds
  • High surface area and capacity for polar analytes
Bond Elut PPL is a styrene-divinylbenzene (SDVB) polymer that has been modified with a proprietary nonpolar surface. PPL will retain even the most polar classes of analytes, including phenols. The large particle size allows ease of flow for viscous or particulate-rich water samples, while the high surface area and strong hydrophobicity ensure reproducible extractions with high recoveries upon elution.