Order Entry
Hungary
ContactUsLinkComponent
Human recombinant Matrix Metalloproteinase-13
Human recombinant Matrix Metalloproteinase-13
Catalog # APOSBITP1340-10UG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant Matrix Metalloproteinase-13
Catalog # APOSBITP1340-10UG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Enzyme type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    Ambient
  • MDL No.:
    MFCD01867476
  • Enzyme name:
    Matrix Metalloproteinase-13
  • Enzyme synonyms:
    MMP-13

Specifications

About this item