Human recombinant Adenosine Deaminase tRNA-Specific 1
Supplier: Apollo Scientific
Certificates
About this item
Specifications
- Enzyme type:Recombinant
- Species:Human
- Storage conditions:Ambient
- Enzyme name:Adenosine Deaminase tRNA-Specific 1
- Enzyme synonyms:ADAT1
Specifications
Frequently Bought Together