Bacteria recombinant malate dehydrogenase
Supplier: Apollo Scientific
Certificates
About this item
Lyophilized powder. Host cell: E. Coli.Specific activity: IU/mg protein>550units. IU per 1mg powder is approximately 400 units.
Specifications
- Enzyme type:Recombinant
- Species:Bacteria
- Storage conditions:Freezer
- MDL No.:MFCD00131567
- Enzyme name:Malate dehydrogenase
- Enzyme synonyms:rMDH, L-Malate: NAD+ oxidoreductase, Malic Dehydrogenase
Specifications
Frequently Bought Together