Order Entry
Hungary
ContactUsLinkComponent
 
Anti-CCL11 Mouse Antibody
  APOSBITP1990-1MG
Anti-CCL11 Mouse Antibody
  APOSBITP1990-1MG

 

  • Antigen name:
    Eotaxin-1
  • Antigen symbol:
    CCL11
  • Host:
    Mouse
  • Reactivity:
    Human
  • Pk:
    1 mg