HT-2 Toxin standard solution
Supplier: Apollo Scientific
About this item
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
2 Options Available Below
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- APOSBIMS115-1ML
- APOSBIMS115-5ML
- Description:
- HT-2 Toxin
- HT-2 Toxin
- CAS No.:
- 26934-87-2
- 26934-87-2
- Formula:
- C₂₂H₃₂O₈
- C₂₂H₃₂O₈
- Flash point:
- 2 °C
- 2 °C
- Storage temperature:
- Freezer
- Freezer
Specifications
Product Family Options
Product Information
- ConcentrationPack typePkAvailabilityPrice
- APOSBIMS115-1MLHT-2 ToxinSpecifications:
More
Specifications
Cat. No.APOSBIMS115-1MLDescriptionHT-2 ToxinCAS No.26934-87-2FormulaC₂₂H₃₂O₈Flash point2 °CStorage temperatureFreezer - APOSBIMS115-5MLHT-2 ToxinSpecifications:
More
Specifications
Cat. No.APOSBIMS115-5MLDescriptionHT-2 ToxinCAS No.26934-87-2FormulaC₂₂H₃₂O₈Flash point2 °CStorage temperatureFreezer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



